Assay Details
Gene Symbol: MOXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015529.3 | NP_056344.2 | 5 - 6 | 948 | 91 | |
XM_017010714.1 | XP_016866203.1 | 5 - 6 | 756 | 91 | ||
XM_017010715.1 | XP_016866204.1 | 5 - 6 | 594 | 91 | ||
GenBank mRNA | AK074879.1 | - | 5 - 6 | 663 | 91 | |
AY007239.1 | - | 4 - 5 | 864 | 91 | ||
AY359094.1 | - | 5 - 6 | 872 | 91 | ||
BC018756.1 | - | 5 - 6 | 907 | 4841330 | 91 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06002113.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06026519.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026519.hg.1 | ENST00000367963.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026519.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06026519.hg.1 | TC06002113.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0600191065.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191065.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191065.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191065.hg.1 | TC0600013190.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600191065.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191065.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191065.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000336749.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000367963.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000392401.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | ENST00000489128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013190.hg.1 | NM_015529.3 | Refseq |
