Assay Details
Gene Symbol: LCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002299.3 | NP_002290.2 | 7 - 8 | 2360 | 141 | |
XM_017004088.1 | XP_016859577.1 | 7 - 8 | 2380 | 141 | ||
GenBank mRNA | X07994.1 | - | 7 - 8 | 2360 | 141 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002374.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
carbohydrate metabolic process
response to nutrient
response to hormone
response to sucrose
response to iron(II) ion
response to nickel cation
response to lead ion
response to drug
response to starvation
response to estrogen
polysaccharide digestion
response to ethanol
glycosyl compound metabolic process
lactase activity
beta-glucosidase activity
transferase activity
glycosylceramidase activity
response to hypoxia
carbohydrate metabolic process
response to nutrient
response to hormone
response to sucrose
response to iron(II) ion
response to nickel cation
response to lead ion
response to drug
response to starvation
response to estrogen
polysaccharide digestion
response to ethanol
glycosyl compound metabolic process
lactase activity
beta-glucosidase activity
transferase activity
glycosylceramidase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02037462.hg.1 | TC02002374.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02037462.hg.1 | ENST00000264162.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02037462.hg.1 | ENST00000455227.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02037462.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014402.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200204993.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200204993.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200204993.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200204993.hg.1 | ENST00000264162.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200204993.hg.1 | ENST00000452974.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200204993.hg.1 | NM_002299.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200204993.hg.1 | TC0200014402.hg.1 | Clariom D TC |
