Assay Details
Gene Symbol: SENP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145204.3 | NP_660205.3 | 1 - 2 | 175 | 72 | |
GenBank mRNA | AK292298.1 | - | 1 - 2 | 177 | 72 | |
BC031411.1 | - | 1 - 2 | 52 | 5195120 | 72 | |
DB033241.1 | - | 1 - 2 | 177 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15000654.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK292298.1, BC031411.1, NM_145204.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15005611.hg.1 | ENST00000340912.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005611.hg.1 | ENST00000544171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005611.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005611.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005611.hg.1 | TC15000654.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005610.hg.1 | ENST00000340912.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005610.hg.1 | ENST00000544171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005610.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005610.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005610.hg.1 | TC15000654.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1500155204.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155204.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155204.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155204.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155204.hg.1 | TC1500007777.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500155204.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155204.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155204.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155204.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155202.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155202.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155202.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155202.hg.1 | TC1500007777.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500155202.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155202.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155202.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155203.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155203.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155203.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155203.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155203.hg.1 | TC1500007777.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500155203.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155203.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155203.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155203.hg.1 | NM_145204.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 127872 | - | - |
