Assay Details
Gene Symbol: CDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254.3 | NP_001245.1 | 1 - 2 | 197 | 73 | |
XM_011525541.1 | XP_011523843.1 | 1 - 2 | 402 | 73 | ||
GenBank mRNA | AF022109.1 | - | 1 - 2 | 73 | 73 | |
AK313620.1 | - | 1 - 2 | 212 | 73 | ||
BC025232.1 | - | 1 - 2 | 176 | 5457941 | 73 | |
U77949.1 | - | 1 - 2 | 197 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000495.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF022109.1,AK313620.1,BC025232.1,NM_001254.3,U77949.1,XM_011525541.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication checkpoint
regulation of cyclin-dependent protein serine/threonine kinase activity
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
DNA replication
DNA replication initiation
mitotic nuclear division
traversing start control point of mitotic cell cycle
negative regulation of DNA replication
negative regulation of cell proliferation
regulation of mitotic metaphase/anaphase transition
positive regulation of cytokinesis
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of fibroblast proliferation
cell division
positive regulation of chromosome segregation
cellular response to vasopressin
cellular response to angiotensin
nucleotide binding
protein binding
ATP binding
kinase binding
DNA replication checkpoint
regulation of cyclin-dependent protein serine/threonine kinase activity
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
DNA replication
DNA replication initiation
mitotic nuclear division
traversing start control point of mitotic cell cycle
negative regulation of DNA replication
negative regulation of cell proliferation
regulation of mitotic metaphase/anaphase transition
positive regulation of cytokinesis
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of fibroblast proliferation
cell division
positive regulation of chromosome segregation
cellular response to vasopressin
cellular response to angiotensin
nucleotide binding
protein binding
ATP binding
kinase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17005961.hg.1 | ENST00000209728.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17005961.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005961.hg.1 | TC17000495.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17005963.hg.1 | ENST00000209728.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17005963.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005963.hg.1 | TC17000495.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000473555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000473555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000473555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000473555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000582402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000582402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000582402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000582402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | ENST00000602403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156662.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156662.hg.1 | ENST00000473555.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156662.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156662.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156662.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156662.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156662.hg.1 | ENST00000473555.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156662.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156662.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156662.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156662.hg.1 | TC1700007790.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700156658.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156658.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156658.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156658.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156658.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156658.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156658.hg.1 | TC1700007790.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700156657.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156657.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156657.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156657.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156657.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156657.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156657.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156657.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156657.hg.1 | TC1700007790.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000209728.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000473555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000473555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000473555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000473555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000577249.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000580824.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000582402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000582402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000582402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000582402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | ENST00000602403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007790.hg.1 | NM_001254.3 | Refseq |
