Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015759.hg.1 |
TC09001226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015759.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015759.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015760.hg.1 |
TC09001226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015760.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015760.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
ENST00000464095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010482.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171719.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171719.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171719.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171719.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171719.hg.1 |
TC0900010482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900171717.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171717.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171717.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171717.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171717.hg.1 |
TC0900010482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900171718.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171718.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171718.hg.1 |
ENST00000341700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171718.hg.1 |
NM_004297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171718.hg.1 |
TC0900010482.hg.1 |
Clariom D TC |