Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037259.hg.1 |
ENST00000319332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037259.hg.1 |
ENST00000427335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037259.hg.1 |
NM_001134418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037259.hg.1 |
NM_018192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037259.hg.1 |
TC03002106.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037258.hg.1 |
ENST00000319332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037258.hg.1 |
ENST00000427335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037258.hg.1 |
NM_001134418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037258.hg.1 |
NM_018192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037258.hg.1 |
TC03002106.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
ENST00000319332.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
ENST00000427335.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
ENST00000463171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
ENST00000490940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
NM_001134418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
NM_018192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
ENST00000463171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
TC0300013513.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
ENST00000319332.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
ENST00000427335.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
ENST00000490940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
NM_001134418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200827.hg.1 |
NM_018192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
ENST00000319332.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
ENST00000427335.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
ENST00000463171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
ENST00000467131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
ENST00000490940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
NM_001134418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
NM_018192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
TC0300013513.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
ENST00000319332.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
ENST00000427335.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
ENST00000463171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
ENST00000467131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
ENST00000490940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
NM_001134418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200828.hg.1 |
NM_018192.3 |
Refseq |