Assay Details
Gene Symbol: MERTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006343.2 | NP_006334.2 | 16 - 17 | 2312 | 93 | |
XM_005263565.4 | XP_005263622.1 | 16 - 17 | 2447 | 93 | ||
XM_011510490.2 | XP_011508792.1 | 16 - 17 | 2347 | 93 | ||
XM_017003164.1 | XP_016858653.1 | 16 - 17 | 2317 | 93 | ||
XM_017003165.1 | XP_016858654.1 | 17 - 18 | 2416 | 93 | ||
GenBank mRNA | AK294196.1 | - | 8 - 9 | 758 | 93 | |
AK308695.1 | - | 8 - 9 | 760 | 93 | ||
AK316584.1 | - | 16 - 17 | 2319 | 93 | ||
BC114917.1 | - | 16 - 17 | 2026 | 40068588 | 93 | |
BC114918.1 | - | 16 - 17 | 2026 | 40012335 | 93 | |
U08023.1 | - | 16 - 17 | 2327 | 93 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000701.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
natural killer cell differentiation
protein phosphorylation
phagocytosis
cell surface receptor signaling pathway
cell-cell signaling
spermatogenesis
peptidyl-tyrosine phosphorylation
platelet activation
secretion by cell
substrate adhesion-dependent cell spreading
apoptotic cell clearance
protein kinase B signaling
positive regulation of phagocytosis
leukocyte migration
negative regulation of lymphocyte activation
retina development in camera-type eye
vagina development
negative regulation of leukocyte apoptotic process
transmembrane receptor protein tyrosine kinase activity
protein binding
ATP binding
natural killer cell differentiation
protein phosphorylation
phagocytosis
cell surface receptor signaling pathway
cell-cell signaling
spermatogenesis
peptidyl-tyrosine phosphorylation
platelet activation
secretion by cell
substrate adhesion-dependent cell spreading
apoptotic cell clearance
protein kinase B signaling
positive regulation of phagocytosis
leukocyte migration
negative regulation of lymphocyte activation
retina development in camera-type eye
vagina development
negative regulation of leukocyte apoptotic process
transmembrane receptor protein tyrosine kinase activity
protein binding
ATP binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000295408.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000409780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000421804.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000439966.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000449344.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000449344.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000449344.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000449344.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000449344.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000449344.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000473065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000473065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000473065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000473065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | ENST00000616902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008894.hg.1 | NM_006343.2 | Refseq |
