Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000317509.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000440567.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000442299.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000451782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000539550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000557907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000557997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000558201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000558379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000558939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000559048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000559929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000560589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
ENST00000560939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
NM_001105192.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
NM_005078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
NM_020908.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015469.hg.1 |
TC15001606.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000317509.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000440567.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000451782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000539550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000557815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000557907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000557919.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000557997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000558201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000558379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000558939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000559048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000559929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000560525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000560589.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000560939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000561453.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000627388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_001105192.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_001282979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_001282980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_001282981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_001282982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_005078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_020908.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
TC1500009904.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000539550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000317509.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000440567.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000451782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000557815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000557907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000557919.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000557997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000558201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000558379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000558939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000559048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000559929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000560525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000560589.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000560939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000561453.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
ENST00000627388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_001105192.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_001282979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_001282980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_001282981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_001282982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_005078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173436.hg.1 |
NM_020908.2 |
Refseq |