Assay Details
Gene Symbol: CAMSAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080429.2 | NP_001073898.1 | 8 - 9 | 1078 | 61 | |
NM_020902.1 | NP_065953.1 | 6 - 7 | 997 | 61 | ||
XM_005272491.3 | XP_005272548.1 | 7 - 8 | 1014 | 61 | ||
XM_011528154.2 | XP_011526456.1 | 8 - 9 | 1048 | 61 | ||
XM_017027036.1 | XP_016882525.1 | 8 - 9 | 1051 | 61 | ||
GenBank mRNA | BC020431.1 | - | 8 - 9 | 921 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000130.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
negative regulation of phosphatase activity
neuron projection development
regulation of organelle organization
microtubule anchoring
zonula adherens maintenance
regulation of microtubule cytoskeleton organization
epithelial cell-cell adhesion
calmodulin binding
spectrin binding
microtubule minus-end binding
microtubule cytoskeleton organization
negative regulation of phosphatase activity
neuron projection development
regulation of organelle organization
microtubule anchoring
zonula adherens maintenance
regulation of microtubule cytoskeleton organization
epithelial cell-cell adhesion
calmodulin binding
spectrin binding
microtubule minus-end binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19002031.hg.1 | ENST00000160298.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19002031.hg.1 | ENST00000446248.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19002031.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19002031.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19002031.hg.1 | TC19000130.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000593434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000593434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000593434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900148296.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900148296.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900148296.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900148296.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900148296.hg.1 | TC1900006848.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900148296.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900148296.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900148296.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900148296.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000160298.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000446248.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000593434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000593434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000593434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | ENST00000595692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_001080429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006848.hg.1 | NM_020902.1 | Refseq |
