Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026520.hg.1 |
TC06002113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026520.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026520.hg.1 |
ENST00000367963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026520.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000392401.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000392401.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000392401.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000489128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000489128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000489128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000489128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
ENST00000489128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000392401.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000392401.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000392401.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000489128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000489128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000489128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000489128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
ENST00000489128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013190.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191066.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191066.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191066.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191066.hg.1 |
ENST00000336749.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191066.hg.1 |
ENST00000367963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191066.hg.1 |
NM_015529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191066.hg.1 |
TC0600013190.hg.1 |
Clariom D TC |