Assay Details
Gene Symbol: ADAMTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014244.4 | NP_055059.2 | 12 - 13 | 2059 | 105 | |
XM_017010063.1 | XP_016865552.1 | 11 - 12 | 1495 | 105 | ||
GenBank mRNA | AJ003125.1 | - | 12 - 13 | 1957 | 105 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05002130.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05029686.hg.1 | ENST00000251582.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029686.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029686.hg.1 | TC05002130.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191273.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191273.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191273.hg.1 | TC0500013039.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500191273.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191273.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
