Assay Details
Gene Symbol: SLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001304743.1 | NP_001291672.1 | 13 - 14 | 3480 | 68 | |
NM_014720.3 | NP_055535.2 | 14 - 15 | 3573 | 68 | ||
XM_011540401.2 | XP_011538703.1 | 13 - 14 | 2229 | 68 | ||
GenBank mRNA | AB002804.1 | - | 13 - 14 | 3151 | 68 | |
AF273048.1 | - | 14 - 15 | 3046 | 68 | ||
AK302855.1 | - | 13 - 14 | 2913 | 68 | ||
BC111565.1 | - | 13 - 14 | 2950 | 40009070 | 68 | |
D86959.1 | - | 13 - 14 | 3430 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000779.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
regulation of cell migration
cytoplasmic microtubule organization
regulation of apoptotic process
protein autophosphorylation
regulation of focal adhesion assembly
cell-cell adhesion
protein serine/threonine kinase activity
protein binding
ATP binding
identical protein binding
protein homodimerization activity
cadherin binding involved in cell-cell adhesion
apoptotic process
regulation of cell migration
cytoplasmic microtubule organization
regulation of apoptotic process
protein autophosphorylation
regulation of focal adhesion assembly
cell-cell adhesion
protein serine/threonine kinase activity
protein binding
ATP binding
identical protein binding
protein homodimerization activity
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10009295.hg.1 | ENST00000335753.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009295.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009295.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10009295.hg.1 | TC10000779.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000160184.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000160184.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000160184.hg.1 | ENST00000474260.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000160184.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000160184.hg.1 | NM_014720.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000160184.hg.1 | TC1000008795.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000160184.hg.1 | NM_001304743.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000160184.hg.1 | ENST00000335753.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000160184.hg.1 | ENST00000369755.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000160184.hg.1 | ENST00000474260.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000160184.hg.1 | NM_014720.3 | Refseq |
