Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006931.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006931.hg.1 |
ENST00000324342.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006931.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006931.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006931.hg.1 |
TC0X000519.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
ENST00000418562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008054.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155061.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155061.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155061.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155061.hg.1 |
NM_194463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155061.hg.1 |
TC0X00008054.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00155061.hg.1 |
ENST00000255499.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155061.hg.1 |
ENST00000324342.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155061.hg.1 |
NM_024539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155061.hg.1 |
NM_194463.1 |
Refseq |