Assay Details
Gene Symbol: ADAM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003812.3 | NP_003803.1 | 7 - 8 | 1016 | 106 | |
XM_005246932.3 | XP_005246989.1 | 7 - 8 | 1116 | 106 | ||
XM_011512086.2 | XP_011510388.1 | 7 - 8 | 1116 | 106 | ||
GenBank mRNA | AB009672.1 | - | 7 - 8 | 1016 | 106 | |
AJ005580.1 | - | 7 - 8 | 845 | 106 | ||
BC132763.1 | - | 7 - 8 | 996 | 40146785 | 106 | |
BC132765.1 | - | 7 - 8 | 996 | 40146787 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001218.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02018679.hg.1 | ENST00000264377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02018679.hg.1 | ENST00000374415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02018679.hg.1 | ENST00000374416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02018679.hg.1 | ENST00000431817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02018679.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02018679.hg.1 | TC02001218.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000444281.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000444281.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000444281.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000444281.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | ENST00000444281.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000444281.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000444281.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000444281.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000444281.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | ENST00000444281.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010577.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200174902.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174902.hg.1 | ENST00000374415.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174902.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200174902.hg.1 | TC0200010577.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200174902.hg.1 | NM_003812.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200174902.hg.1 | ENST00000264377.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174902.hg.1 | ENST00000374415.7 | Ensembl |
