Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004849.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004849.hg.1 |
ENST00000539142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004849.hg.1 |
ENST00000551171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004849.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004849.hg.1 |
TC14000410.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000547463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000547463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000547463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000552873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000552873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
ENST00000552873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152651.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152651.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152651.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152651.hg.1 |
TC1400007504.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400152651.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152651.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152651.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152652.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152652.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152652.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152652.hg.1 |
TC1400007504.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400152652.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152652.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152652.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000267502.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000547463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000547463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000547463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000551171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000552873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000552873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
ENST00000552873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007504.hg.1 |
NM_152443.2 |
Refseq |