Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006713.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006713.hg.1 |
ENST00000394142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006713.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006713.hg.1 |
TC06000588.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0600156006.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156006.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156006.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156006.hg.1 |
TC0600008059.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600156006.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156006.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156006.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000372836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
ENST00000394142.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008059.hg.1 |
NM_006586.4 |
Refseq |