Assay Details
Gene Symbol: CCNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017013958.1 | XP_016869447.1 | 1 - 2 | 215 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017013958.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle checkpoint signaling
regulation of cyclin-dependent protein serine/threonine kinase activity
G1/S transition of mitotic cell cycle
DNA replication initiation
cell division
protein binding
cyclin-dependent protein serine/threonine kinase regulator activity
protein kinase binding
cell cycle checkpoint signaling
regulation of cyclin-dependent protein serine/threonine kinase activity
G1/S transition of mitotic cell cycle
DNA replication initiation
cell division
protein binding
cyclin-dependent protein serine/threonine kinase regulator activity
protein kinase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000517487.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000517487.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000517487.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000517487.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000517487.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000524224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000524224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000524224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000524224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000524224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000308108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000396133.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000517487.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000517487.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000517487.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000517487.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000517487.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000519889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000520509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000521809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000523476.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000524224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000524224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000524224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000524224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000524224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | ENST00000611101.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011148.hg.1 | NM_057749.2 | Refseq |
