Assay Details
Gene Symbol: BAIAP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025045.5 | NP_079321.3 | 6 - 7 | 609 | 56 | |
XM_005261751.4 | XP_005261808.1 | 7 - 8 | 610 | 56 | ||
XM_011530379.2 | XP_011528681.1 | 6 - 7 | 726 | 56 | ||
XM_011530380.2 | XP_011528682.1 | 5 - 6 | 512 | 56 | ||
XM_011530381.2 | XP_011528683.1 | 6 - 7 | 503 | 56 | ||
XM_011530382.2 | XP_011528684.1 | 6 - 7 | 891 | 56 | ||
XM_011530383.2 | XP_011528685.1 | 5 - 6 | 829 | 56 | ||
XM_011530384.2 | XP_011528686.1 | 6 - 7 | 767 | 56 | ||
GenBank mRNA | AK026235.1 | - | 6 - 7 | 567 | 56 | |
AY358164.1 | - | 6 - 7 | 609 | 56 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000785.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
plasma membrane organization
insulin receptor signaling pathway
positive regulation of actin filament polymerization
actin filament bundle assembly
actin crosslink formation
membrane organization
positive regulation of actin cytoskeleton reorganization
phospholipid binding
cytoskeletal anchor activity
plasma membrane organization
insulin receptor signaling pathway
positive regulation of actin filament polymerization
actin filament bundle assembly
actin crosslink formation
membrane organization
positive regulation of actin cytoskeleton reorganization
phospholipid binding
cytoskeletal anchor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22013096.hg.1 | TC22000785.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22013096.hg.1 | ENST00000332536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22013096.hg.1 | ENST00000381669.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22013096.hg.1 | ENST00000402500.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22013096.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162730.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162730.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162730.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162730.hg.1 | TC2200008680.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200162730.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162730.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162730.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000332536.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000381669.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | ENST00000428572.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008680.hg.1 | NM_025045.5 | Refseq |
