Assay Details
Gene Symbol: FBXW8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153348.2 | NP_699179.2 | 2 - 3 | 500 | 139 | |
XM_005253867.2 | XP_005253924.1 | 2 - 3 | 447 | 139 | ||
GenBank mRNA | BC037296.2 | - | 2 - 3 | 500 | 4821240 | 139 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000920.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_005253867.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12011962.hg.1 | ENST00000309909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12011962.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12011962.hg.1 | ENST00000505227.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12011962.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12011962.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12011962.hg.1 | TC12000920.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200166907.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200166907.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200166907.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200166907.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200166907.hg.1 | TC1200009036.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200166907.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200166907.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200166907.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200166907.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
