Assay Details
Gene Symbol: FBXW8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012174.1 | NP_036306.1 | 5 - 6 | 716 | 78 | |
NM_153348.2 | NP_699179.2 | 5 - 6 | 914 | 78 | ||
XM_005253867.2 | XP_005253924.1 | 4 - 5 | 772 | 78 | ||
XM_005253868.3 | XP_005253925.1 | 4 - 5 | 605 | 78 | ||
XM_017019175.1 | XP_016874664.1 | 5 - 6 | 691 | 78 | ||
XM_017019176.1 | XP_016874665.1 | 6 - 7 | 923 | 78 | ||
XM_017019177.1 | XP_016874666.1 | 6 - 7 | 923 | 78 | ||
GenBank mRNA | AK289621.1 | - | 5 - 6 | 707 | 78 | |
BC037296.2 | - | 5 - 6 | 914 | 4821240 | 78 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000920.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
