Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000466695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000466695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000466695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000473631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000473631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
ENST00000473631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012735.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012735.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012735.hg.1 |
TC06001077.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012736.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012736.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012736.hg.1 |
TC06001077.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0600167010.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167010.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167010.hg.1 |
TC0600009783.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600167010.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167010.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167011.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167011.hg.1 |
ENST00000473631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167011.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167011.hg.1 |
TC0600009783.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600167011.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167011.hg.1 |
ENST00000473631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167011.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000367463.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000466695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000466695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000466695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000473631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000473631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
ENST00000473631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009783.hg.1 |
NM_005715.2 |
Refseq |