Assay Details
Gene Symbol: ALG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142364.1 | NP_001135836.1 | 1 - 2 | 128 | 119 | |
NM_013338.4 | NP_037470.1 | 1 - 2 | 128 | 119 | ||
XM_017020552.1 | XP_016876041.1 | 1 - 2 | 113 | 119 | ||
GenBank mRNA | AF102850.1 | - | 1 - 2 | 136 | 119 | |
AF161498.1 | - | 1 - 2 | 77 | 119 | ||
AF183413.1 | - | 1 - 2 | 87 | 119 | ||
AK299085.1 | - | 1 - 2 | 104 | 119 | ||
AK299126.1 | - | 1 - 2 | 97 | 119 | ||
BC012531.1 | - | 1 - 2 | 80 | 4273297 | 119 | |
BG482604.1 | - | 1 - 2 | 118 | |||
BP246765.1 | - | 1 - 2 | 128 | 119 | ||
CB988638.1 | - | 125 | 119 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000570.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF102850.1, AF161498.1, AF183413.1, AK299085.1, AK299126.1, BC012531.1, NM_001142364.1, NM_013338.4, XM_017020552.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
protein N-linked glycosylation
GPI anchor biosynthetic process
determination of left/right symmetry
protein N-linked glycosylation via asparagine
dolichol metabolic process
protein O-linked mannosylation
dolichyl-phosphate-mannose-protein mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate beta-glucosyltransferase activity
dolichyl-phosphate beta-D-mannosyltransferase activity
protein glycosylation
protein N-linked glycosylation
GPI anchor biosynthetic process
determination of left/right symmetry
protein N-linked glycosylation via asparagine
dolichol metabolic process
protein O-linked mannosylation
dolichyl-phosphate-mannose-protein mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate beta-glucosyltransferase activity
dolichyl-phosphate beta-D-mannosyltransferase activity
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13006485.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006485.hg.1 | ENST00000413537.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006485.hg.1 | ENST00000443765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006485.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006485.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006485.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006485.hg.1 | TC13000570.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13006488.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006488.hg.1 | ENST00000443765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006488.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006488.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006488.hg.1 | TC13000570.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13006486.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006486.hg.1 | ENST00000413537.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006486.hg.1 | ENST00000443765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006486.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006486.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006486.hg.1 | TC13000570.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158245.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158245.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158245.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158245.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158245.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158245.hg.1 | TC1300008632.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300158245.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158245.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158245.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158245.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158245.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158246.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158246.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158246.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158246.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158246.hg.1 | TC1300008632.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300158246.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158246.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158246.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158246.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158248.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158248.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158248.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158248.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158248.hg.1 | TC1300008632.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300158248.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158248.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158248.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158248.hg.1 | NM_013338.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 129788 | - | - |
