Assay Details
Gene Symbol: CXCL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100812.1 | NP_001094282.1 | 1 - 2 | 664 | 53 | |
NM_022059.3 | NP_071342.2 | 1 - 2 | 664 | 53 | ||
GenBank mRNA | AF275260.1 | - | 1 - 2 | 175 | 53 | |
AF301016.1 | - | 1 - 2 | 555 | 53 | ||
AF337812.1 | - | 1 - 2 | 89 | 53 | ||
AK027389.1 | - | 1 - 2 | 582 | 53 | ||
AK292114.1 | - | 1 - 2 | 506 | 53 | ||
AK315017.1 | - | 1 - 2 | 664 | 53 | ||
AY358909.1 | - | 1 - 2 | 509 | 53 | ||
BC017588.1 | - | 1 - 2 | 658 | 4837010 | 53 | |
BC044930.1 | - | 1 - 2 | 359 | 5762458 | 53 | |
JC209418.1 | - | 1 - 2 | 664 | 53 | ||
JC209420.1 | - | 1 - 2 | 664 | 53 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001029.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
receptor-mediated endocytosis
chemotaxis
T cell chemotaxis
positive regulation of cell growth
positive regulation of cell migration
response to cytokine
response to interferon-gamma
response to tumor necrosis factor
cellular response to lipopolysaccharide
low-density lipoprotein particle receptor activity
scavenger receptor activity
chemokine activity
receptor-mediated endocytosis
chemotaxis
T cell chemotaxis
positive regulation of cell growth
positive regulation of cell migration
response to cytokine
response to interferon-gamma
response to tumor necrosis factor
cellular response to lipopolysaccharide
low-density lipoprotein particle receptor activity
scavenger receptor activity
chemokine activity
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17013886.hg.1 | ENST00000293778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17013886.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17013886.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17013886.hg.1 | TC17001029.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17013887.hg.1 | ENST00000293778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17013887.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17013887.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17013887.hg.1 | TC17001029.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17013885.hg.1 | ENST00000293778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17013885.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17013885.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17013885.hg.1 | TC17001029.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000573123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000573123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000573123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000573123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000575168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000575168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000575168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000575168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000575168.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173328.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173328.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173328.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173328.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173328.hg.1 | TC1700009528.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700173328.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173328.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173328.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173328.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173330.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173330.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173330.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173330.hg.1 | TC1700009528.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700173330.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173330.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173330.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173325.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173325.hg.1 | ENST00000573123.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173325.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173325.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173325.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173325.hg.1 | TC1700009528.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700173325.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173325.hg.1 | ENST00000573123.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173325.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173325.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173325.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173327.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173327.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173327.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173327.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173327.hg.1 | TC1700009528.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700173327.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173327.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173327.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173327.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173329.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173329.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173329.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173329.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173329.hg.1 | TC1700009528.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700173329.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173329.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173329.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173329.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000293778.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000573123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000573123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000573123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000573123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000574412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000575168.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000575168.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000575168.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000575168.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000575168.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | ENST00000576153.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_001100812.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009528.hg.1 | NM_022059.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 199059 | - | - |
