Assay Details
Gene Symbol: TCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001062.3 | NP_001053.2 | 3 - 4 | 498 | 98 | |
GenBank mRNA | AK292990.1 | - | 3 - 4 | 415 | 98 | |
BC018632.1 | - | 3 - 4 | 420 | 98 | ||
BP370727.1 | - | 3 - 4 | 504 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001837.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000529251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000529251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000529251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000529251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000533734.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000533734.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000533734.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11021116.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11021116.hg.1 | ENST00000532419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11021116.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11021116.hg.1 | TC11001837.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100182891.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182891.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182891.hg.1 | ENST00000533734.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182891.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182891.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182891.hg.1 | TC1100010992.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182891.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182891.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182891.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182891.hg.1 | ENST00000533734.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182891.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000529251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000529251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000529251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000529251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000533734.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000533734.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000533734.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010992.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182893.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182893.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182893.hg.1 | ENST00000533734.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182893.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182893.hg.1 | NM_001062.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182893.hg.1 | TC1100010992.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182893.hg.1 | ENST00000257264.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182893.hg.1 | ENST00000532419.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182893.hg.1 | ENST00000533734.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182893.hg.1 | ENST00000534531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182893.hg.1 | NM_001062.3 | Refseq |
