Assay Details
Gene Symbol: RPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002947.4 | NP_002938.1 | 4 - 5 | 418 | 155 | |
GenBank mRNA | BC009868.2 | - | 4 - 5 | 418 | 3940438 | 155 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001139.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
telomere maintenance via recombination
double-strand break repair via homologous recombination
DNA replication
transcription-coupled nucleotide-excision repair
base-excision repair
nucleotide-excision repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
nucleotide-excision repair, DNA gap filling
mismatch repair
regulation of mitotic cell cycle
translesion synthesis
nucleotide-excision repair, DNA incision
interstrand cross-link repair
regulation of cell population proliferation
error-prone translesion synthesis
DNA damage response, detection of DNA damage
error-free translesion synthesis
regulation of cellular response to heat
regulation of signal transduction by p53 class mediator
damaged DNA binding
single-stranded DNA binding
protein binding
G1/S transition of mitotic cell cycle
telomere maintenance via recombination
double-strand break repair via homologous recombination
DNA replication
transcription-coupled nucleotide-excision repair
base-excision repair
nucleotide-excision repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
nucleotide-excision repair, DNA gap filling
mismatch repair
regulation of mitotic cell cycle
translesion synthesis
nucleotide-excision repair, DNA incision
interstrand cross-link repair
regulation of cell population proliferation
error-prone translesion synthesis
DNA damage response, detection of DNA damage
error-free translesion synthesis
regulation of cellular response to heat
regulation of signal transduction by p53 class mediator
damaged DNA binding
single-stranded DNA binding
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07017795.hg.1 | ENST00000223129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017795.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07017795.hg.1 | TC07001139.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07017791.hg.1 | ENST00000223129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017791.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07017791.hg.1 | TC07001139.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07017793.hg.1 | ENST00000223129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017793.hg.1 | ENST00000396682.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017793.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07017793.hg.1 | TC07001139.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07017792.hg.1 | ENST00000223129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017792.hg.1 | ENST00000396682.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017792.hg.1 | ENST00000406109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07017792.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07017792.hg.1 | TC07001139.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000401447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000401447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000401447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000401447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000483031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000483031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | ENST00000483031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172930.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172930.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172930.hg.1 | TC0700010247.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700172930.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172930.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172927.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172927.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172927.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172927.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172927.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172927.hg.1 | TC0700010247.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700172927.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172927.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172927.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172927.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172927.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172928.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172928.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172928.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172928.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172928.hg.1 | TC0700010247.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700172928.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172928.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172928.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172928.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000396682.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000401447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000401447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000401447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000401447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000406109.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000462723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000463632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000483031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000483031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | ENST00000483031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010247.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172926.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172926.hg.1 | NM_002947.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700172926.hg.1 | TC0700010247.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700172926.hg.1 | ENST00000223129.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700172926.hg.1 | NM_002947.4 | Refseq |
