Assay Details
Gene Symbol: CWH43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286791.1 | NP_001273720.1 | 14 - 15 | 1966 | 82 | |
NM_025087.2 | NP_079363.2 | 14 - 15 | 2047 | 82 | ||
XM_011513755.1 | XP_011512057.1 | 14 - 15 | 2105 | 82 | ||
XM_011513756.2 | XP_011512058.1 | 14 - 15 | 2136 | 82 | ||
XM_011513757.2 | XP_011512059.1 | 14 - 15 | 2162 | 82 | ||
XM_011513758.1 | XP_011512060.1 | 13 - 14 | 1847 | 82 | ||
XM_011513759.1 | XP_011512061.1 | 11 - 12 | 1501 | 82 | ||
GenBank mRNA | AI377551.1 | - | 2 - 3 | 82 | ||
AK025164.1 | - | 14 - 15 | 2047 | 82 | ||
AK300495.1 | - | 14 - 15 | 1969 | 82 | ||
AK316307.1 | - | 14 - 15 | 1939 | 82 | ||
BC137387.1 | - | 14 - 15 | 2030 | 9021385 | 82 | |
BC137388.1 | - | 14 - 15 | 2030 | 9021386 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000282.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
