Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021854.hg.1 |
TC06001809.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021854.hg.1 |
ENST00000350082.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021854.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021854.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021854.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183054.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183054.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183054.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183054.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183054.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183054.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183054.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183054.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183054.hg.1 |
TC0600012061.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000350082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
ENST00000356971.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_014920.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012061.hg.1 |
NM_016513.4 |
Refseq |