Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011815.hg.1 |
TC03000658.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011815.hg.1 |
ENST00000319340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011815.hg.1 |
ENST00000383575.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011815.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011819.hg.1 |
TC03000658.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011819.hg.1 |
ENST00000319340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011819.hg.1 |
ENST00000383575.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011819.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011817.hg.1 |
TC03000658.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011817.hg.1 |
ENST00000319340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011817.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161604.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161604.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161604.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161604.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161604.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161604.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161604.hg.1 |
TC0300008677.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008677.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161608.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161608.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161608.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161608.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161608.hg.1 |
ENST00000615522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161608.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161608.hg.1 |
TC0300008677.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300161606.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161606.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161606.hg.1 |
ENST00000319340.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161606.hg.1 |
NM_152889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161606.hg.1 |
TC0300008677.hg.1 |
Clariom D TC |