Assay Details
Gene Symbol: CCDC186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321829.1 | NP_001308758.1 | 1 - 2 | 204 | 84 | |
NM_153249.1 | NP_694981.1 | 1 - 2 | 204 | 84 | ||
NR_135815.1 | 1 - 2 | 204 | 84 | |||
GenBank mRNA | AK092620.1 | - | 1 - 2 | 187 | 84 | |
BC100670.1 | - | 1 - 2 | 131 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK092620.1, NM_001321829.1, NM_153249.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000459421.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000459421.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000497592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000497592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NR_031747.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NR_031747.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000459421.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000459421.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000497592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000497592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NR_031747.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NR_031747.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 124534 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 185732 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 221878 | - | - |
