Assay Details
Gene Symbol: LPCAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024830.3 | NP_079106.3 | 4 - 5 | 721 | 70 | |
XM_005248373.3 | XP_005248430.1 | 5 - 6 | 629 | 70 | ||
XM_011514132.1 | XP_011512434.1 | 4 - 5 | 730 | 70 | ||
XM_011514134.1 | XP_011512436.1 | 4 - 5 | 496 | 70 | ||
GenBank mRNA | AB244719.2 | - | 4 - 5 | 604 | 70 | |
AK090444.1 | - | 2 - 3 | 200 | 70 | ||
BC020166.1 | - | 2 - 3 | 187 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001110.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidic acid biosynthetic process
phospholipid biosynthetic process
triglyceride biosynthetic process
phosphatidylglycerol acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
surfactant homeostasis
positive regulation of protein catabolic process
retina development in camera-type eye
negative regulation of phosphatidylcholine biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity
calcium ion binding
2-acylglycerol-3-phosphate O-acyltransferase activity
1-alkenylglycerophosphocholine O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acetyltransferase activity
phosphatidic acid biosynthetic process
phospholipid biosynthetic process
triglyceride biosynthetic process
phosphatidylglycerol acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
surfactant homeostasis
positive regulation of protein catabolic process
retina development in camera-type eye
negative regulation of phosphatidylcholine biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity
calcium ion binding
2-acylglycerol-3-phosphate O-acyltransferase activity
1-alkenylglycerophosphocholine O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acetyltransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05016192.hg.1 | TC05001110.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05016192.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05016192.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | ENST00000507282.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | ENST00000507282.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500169655.hg.1 | TC0500009863.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
