Assay Details
Gene Symbol: TRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX538160.1 | - | 1 - 2 | 41 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 199702 | - | - |
