Assay Details
Gene Symbol: USP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032410.1 | NP_001027582.1 | 2 - 3 | 216 | 122 | |
XM_017028258.1 | XP_016883747.1 | 2 - 3 | 182 | 122 | ||
XM_017028261.1 | XP_016883750.1 | 2 - 3 | 157 | 122 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001032410.1, XM_017028258.1, XM_017028261.1 transcripts.
This assay is in the 5'UTR region of the NM_001032410.1, XM_017028258.1, XM_017028261.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
