Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002913.hg.1 |
TC6_qbl_hap6000153.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002913.hg.1 |
NM_001098534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002913.hg.1 |
NM_001199697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002913.hg.1 |
NM_001199698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002913.hg.1 |
NM_004639.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002913.hg.1 |
NM_080702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002913.hg.1 |
NM_080703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002459.hg.1 |
TC6_mann_hap4000138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002459.hg.1 |
NM_001098534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002459.hg.1 |
NM_001199697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002459.hg.1 |
NM_001199698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002459.hg.1 |
NM_004639.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002459.hg.1 |
NM_080702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002459.hg.1 |
NM_080703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002600.hg.1 |
TC6_mcf_hap5000138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002600.hg.1 |
NM_001098534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002600.hg.1 |
NM_001199697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002600.hg.1 |
NM_001199698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002600.hg.1 |
NM_004639.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002600.hg.1 |
NM_080702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002600.hg.1 |
NM_080703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002603.hg.1 |
TC6_ssto_hap7000133.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002603.hg.1 |
NM_001098534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002603.hg.1 |
NM_001199697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002603.hg.1 |
NM_001199698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002603.hg.1 |
NM_080703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002603.hg.1 |
NM_004639.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002603.hg.1 |
NM_080702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
ENST00000211379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
ENST00000362049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
ENST00000375964.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
ENST00000375976.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
ENST00000404765.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
ENST00000439687.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
NM_001098534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
NM_001199697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
NM_001199698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
NM_004639.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
NM_080702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
NM_080703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017603.hg.1 |
TC06001532.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002933.hg.1 |
TC6_cox_hap2000161.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002933.hg.1 |
NM_001098534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002933.hg.1 |
NM_001199697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002933.hg.1 |
NM_001199698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002933.hg.1 |
NM_004639.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002933.hg.1 |
NM_080702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002933.hg.1 |
NM_080703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002843.hg.1 |
TC6_dbb_hap3000150.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002843.hg.1 |
NM_001098534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002843.hg.1 |
NM_001199697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002843.hg.1 |
NM_001199698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002843.hg.1 |
NM_004639.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002843.hg.1 |
NM_080702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002843.hg.1 |
NM_080703.2 |
Refseq |