Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006008.hg.1 |
ENST00000343932.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006008.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006008.hg.1 |
TC15000684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006007.hg.1 |
ENST00000343932.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006007.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006007.hg.1 |
TC15000684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007855.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156031.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156031.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156031.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156031.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156031.hg.1 |
TC1500007855.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500156029.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156029.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156029.hg.1 |
ENST00000343932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156029.hg.1 |
NM_000761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156029.hg.1 |
TC1500007855.hg.1 |
Clariom D TC |