Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000391.hg.1 |
ENST00000263126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000391.hg.1 |
ENST00000380448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000391.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000391.hg.1 |
TC10000044.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000469875.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000469875.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000469875.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000469875.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
ENST00000469875.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012517.hg.1 |
ENST00000427165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012517.hg.1 |
ENST00000427165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012517.hg.1 |
ENST00000427165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012517.hg.1 |
ENST00000427165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012517.hg.1 |
ENST00000427165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000392.hg.1 |
ENST00000263126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000392.hg.1 |
ENST00000380448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000392.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000392.hg.1 |
TC10000044.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000469875.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000469875.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000469875.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000469875.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
ENST00000469875.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006611.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145293.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145293.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145293.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145293.hg.1 |
TC1000006611.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000145293.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145293.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145293.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145294.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145294.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145294.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145294.hg.1 |
TC1000006611.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000145294.hg.1 |
ENST00000263126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145294.hg.1 |
ENST00000380448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145294.hg.1 |
NM_001818.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012517.hg.1 |
ENST00000427165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012517.hg.1 |
ENST00000427165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012517.hg.1 |
ENST00000427165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012517.hg.1 |
ENST00000427165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012517.hg.1 |
ENST00000427165.1 |
Ensembl |