Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000628457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000628457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180280.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180280.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180280.hg.1 |
TC0400011693.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400180280.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180280.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180292.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180292.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180292.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180292.hg.1 |
TC0400011693.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400180292.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180292.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180292.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000628457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000628457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011693.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180285.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180285.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180285.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180285.hg.1 |
TC0400011693.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400180285.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180285.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180285.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180290.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180290.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180290.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180290.hg.1 |
TC0400011693.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400180290.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180290.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180290.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180294.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180294.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180294.hg.1 |
NR_125930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180294.hg.1 |
TC0400011693.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400180294.hg.1 |
ENST00000629093.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180294.hg.1 |
ENST00000629462.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180294.hg.1 |
NR_125930.1 |
Refseq |