Assay Details
Gene Symbol: E2F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001949.4 | NP_001940.1 | 2 - 3 | 829 | 88 | |
XM_005248865.4 | XP_005248922.1 | 2 - 3 | 323 | 88 | ||
XM_005248866.4 | XP_005248923.1 | 2 - 3 | 1052 | 88 | ||
XM_011514328.2 | XP_011512630.1 | 2 - 3 | 1085 | 88 | ||
XM_017010330.1 | XP_016865819.1 | 2 - 3 | 1108 | 88 | ||
GenBank mRNA | Y10479.1 | - | 2 - 3 | 568 | 88 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000128.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription initiation from RNA polymerase II promoter
cell cycle
positive regulation of cell population proliferation
positive regulation of transcription, DNA-templated
negative regulation of fat cell proliferation
core promoter sequence-specific DNA binding
DNA-binding transcription factor activity
protein binding
transcription initiation from RNA polymerase II promoter
cell cycle
positive regulation of cell population proliferation
positive regulation of transcription, DNA-templated
negative regulation of fat cell proliferation
core promoter sequence-specific DNA binding
DNA-binding transcription factor activity
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06001305.hg.1 | ENST00000346618.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06001305.hg.1 | ENST00000535432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06001305.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06001305.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06001305.hg.1 | TC06000128.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06001304.hg.1 | ENST00000346618.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06001304.hg.1 | ENST00000378646.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06001304.hg.1 | ENST00000535432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06001304.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06001304.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06001304.hg.1 | TC06000128.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0600148568.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600148568.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600148568.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600148568.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600148568.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600148568.hg.1 | TC0600007138.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600148568.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600148568.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600148568.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600148568.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600148568.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
