Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004494.hg.1 |
ENST00000252506.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004494.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004494.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004494.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004494.hg.1 |
TC09000418.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004495.hg.1 |
ENST00000252506.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004495.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004495.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004495.hg.1 |
TC09000418.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900153111.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153111.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153111.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153111.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153111.hg.1 |
TC0900007877.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900153111.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153111.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153111.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153111.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
ENST00000494726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007877.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153113.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153113.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153113.hg.1 |
NM_006705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153113.hg.1 |
TC0900007877.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900153113.hg.1 |
ENST00000252506.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153113.hg.1 |
ENST00000375769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153113.hg.1 |
NM_006705.3 |
Refseq |