Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006725.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006725.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006725.hg.1 |
TC06000590.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0600156025.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156025.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156025.hg.1 |
TC0600008062.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600156025.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156025.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
ENST00000372808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008062.hg.1 |
NM_018960.5 |
Refseq |