Assay Details
Gene Symbol: EPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015630.3 | NP_056445.3 | 3 - 4 | 489 | 114 | |
XM_011510941.2 | XP_011509243.1 | 3 - 4 | 700 | 114 | ||
XM_011510943.2 | XP_011509245.1 | 2 - 3 | 373 | 114 | ||
GenBank mRNA | AF286904.1 | - | 3 - 4 | 481 | 114 | |
AK001433.1 | - | 1 - 2 | 80 | 114 | ||
AK125789.1 | - | 4 - 5 | 558 | 114 | ||
BC093818.1 | - | 3 - 4 | 401 | 7939663 | 114 | |
BC093820.1 | - | 3 - 4 | 401 | 7939665 | 114 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000926.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK001433.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02013515.hg.1 | ENST00000258484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013515.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02013515.hg.1 | TC02000926.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02013514.hg.1 | ENST00000258484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013514.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02013514.hg.1 | TC02000926.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0200167197.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167197.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167197.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167197.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167197.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167197.hg.1 | TC0200009612.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200167197.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167197.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167197.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167197.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167197.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167198.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167198.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167198.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167198.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200167198.hg.1 | TC0200009612.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200167198.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167198.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167198.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200167198.hg.1 | NM_015630.3 | Refseq |
