Assay Details
Gene Symbol: KPNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012316.4 | NP_036448.1 | 3 - 4 | 320 | 86 | |
XM_005270711.3 | XP_005270768.1 | 3 - 4 | 681 | 86 | ||
GenBank mRNA | AB621806.1 | - | 3 - 4 | 309 | 86 | |
AF060543.1 | - | 3 - 4 | 287 | 86 | ||
AK002111.1 | - | 3 - 4 | 307 | 86 | ||
AK295615.1 | - | 3 - 4 | 435 | 86 | ||
AK301714.1 | - | 3 - 4 | 274 | 86 | ||
AK315490.1 | - | 3 - 4 | 307 | 86 | ||
BC020520.1 | - | 3 - 4 | 295 | 3913864 | 86 | |
BP374991.1 | - | 3 - 4 | 320 | 86 | ||
BT009843.1 | - | 2 - 3 | 232 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000419.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01006396.hg.1 | ENST00000373617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01006396.hg.1 | ENST00000373625.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01006396.hg.1 | ENST00000471599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01006396.hg.1 | ENST00000537234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01006396.hg.1 | ENST00000545542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01006396.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01006396.hg.1 | TC01000419.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0100153793.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153793.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153793.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153793.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153793.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100153793.hg.1 | TC0100007662.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100153793.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153793.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153793.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153793.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153793.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
