Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004283.hg.1 |
ENST00000288207.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004283.hg.1 |
ENST00000559622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004283.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004283.hg.1 |
TC15000449.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000561077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000561077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000561077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000561077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000561077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000561077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000561077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000561077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000561077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000561077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007392.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500152650.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152650.hg.1 |
ENST00000559301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152650.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152650.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152650.hg.1 |
NM_004701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500152650.hg.1 |
ENST00000621385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152650.hg.1 |
TC1500007392.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500152650.hg.1 |
ENST00000288207.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152650.hg.1 |
ENST00000559301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152650.hg.1 |
ENST00000559622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152650.hg.1 |
NM_004701.3 |
Refseq |