Assay Details
Gene Symbol: CHRNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199279.1 | NP_001186208.1 | 3 - 4 | 684 | 69 | |
NM_004198.3 | NP_004189.1 | 4 - 5 | 729 | 69 | ||
GenBank mRNA | AK298798.1 | - | 3 - 4 | 508 | 69 | |
AK313521.1 | - | 4 - 5 | 575 | 69 | ||
BC014456.1 | - | 4 - 5 | 419 | 4862198 | 69 | |
U62435.1 | - | 4 - 5 | 515 | 69 | ||
Y16282.1 | - | 4 - 5 | 373 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001182.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transport
signal transduction
chemical synaptic transmission
synaptic transmission, cholinergic
neuromuscular synaptic transmission
regulation of dopamine secretion
response to nicotine
membrane depolarization
cation transmembrane transport
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine-gated cation-selective channel activity
acetylcholine binding
transport
signal transduction
chemical synaptic transmission
synaptic transmission, cholinergic
neuromuscular synaptic transmission
regulation of dopamine secretion
response to nicotine
membrane depolarization
cation transmembrane transport
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine-gated cation-selective channel activity
acetylcholine binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08015240.hg.1 | ENST00000276410.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08015240.hg.1 | ENST00000534622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08015240.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08015240.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08015240.hg.1 | TC08001182.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800170037.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170037.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170037.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800170037.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800170037.hg.1 | TC0800010297.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800170037.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800170037.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170037.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170037.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800170038.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170038.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170038.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170038.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800170038.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800170038.hg.1 | TC0800010297.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800170038.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170038.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170038.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170038.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800170038.hg.1 | NM_004198.3 | Refseq |
