Assay Details
Gene Symbol: ANKMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020319.2 | NP_064715.1 | 3 - 4 | 512 | 78 | |
XM_011515458.1 | XP_011513760.1 | 4 - 5 | 639 | 78 | ||
GenBank mRNA | AK001389.1 | - | 3 - 4 | 481 | 78 | |
AK001740.1 | - | 3 - 4 | 450 | 78 | ||
AK023704.1 | - | 3 - 4 | 449 | 78 | ||
BC035353.1 | - | 3 - 4 | 468 | 5185501 | 78 | |
DA210027.1 | - | 3 - 4 | 513 | 78 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001161.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07018174.hg.1 | TC07001161.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07018174.hg.1 | ENST00000306999.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018174.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07018175.hg.1 | TC07001161.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07018175.hg.1 | ENST00000306999.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018175.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173584.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173584.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173584.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173584.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173584.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173584.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173584.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173584.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173584.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173584.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173584.hg.1 | TC0700010352.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173585.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173585.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173585.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173585.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173585.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173585.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173585.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173585.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173585.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173585.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173585.hg.1 | TC0700010352.hg.1 | Clariom D TC |
