Assay Details
Gene Symbol: DOC2GP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033791.1 | 1 - 2 | 49 | 74 | ||
GenBank mRNA | DY654453.1 | - | 3 - 4 | 302 | 74 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001990.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11023927.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11023927.hg.1 | TC11001990.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011344.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11023926.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11023926.hg.1 | TC11001990.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11023928.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11023928.hg.1 | TC11001990.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100187509.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187509.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187509.hg.1 | TC1100011344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100187509.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187509.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187508.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187508.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187508.hg.1 | TC1100011344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100187508.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187508.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187510.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187510.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187510.hg.1 | TC1100011344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100187510.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187510.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000495263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | ENST00000514950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | NR_033791.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011344.hg.1 | NR_033791.1 | Refseq |
