Assay Details
Gene Symbol: GAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143830.1 | NP_001137302.1 | 4 - 5 | 720 | 82 | |
NM_005256.3 | NP_005247.1 | 3 - 4 | 511 | 82 | ||
NM_177553.2 | NP_808221.1 | 4 - 5 | 662 | 82 | ||
XM_011519971.2 | XP_011518273.1 | 4 - 5 | 720 | 82 | ||
XM_011519972.2 | XP_011518274.1 | 4 - 5 | 2715 | 82 | ||
XM_011519975.1 | XP_011518277.1 | 4 - 5 | 720 | 82 | ||
XM_011519978.2 | XP_011518280.1 | 4 - 5 | 720 | 82 | ||
XM_017017528.1 | XP_016873017.1 | 4 - 5 | 1412 | 82 | ||
XM_017017529.1 | XP_016873018.1 | 4 - 5 | 557 | 82 | ||
XM_017017530.1 | XP_016873019.1 | 4 - 5 | 750 | 82 | ||
XM_017017531.1 | XP_016873020.1 | 4 - 5 | 571 | 82 | ||
XM_017017532.1 | XP_016873021.1 | 4 - 5 | 720 | 82 | ||
GenBank mRNA | AK313734.1 | - | 4 - 5 | 726 | 82 | |
BC040470.1 | - | 4 - 5 | 662 | 4280621 | 82 | |
CR450285.1 | - | 3 - 4 | 414 | 82 | ||
U95032.1 | - | 3 - 4 | 480 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000265.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11003562.hg.1 | ENST00000278187.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003562.hg.1 | ENST00000433790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003562.hg.1 | ENST00000454584.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003562.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003562.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003562.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003562.hg.1 | TC11000265.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | TC1100007069.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149920.hg.1 | NM_177553.2 | Refseq |
