Assay Details
Gene Symbol: TAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152787.4 | NP_690000.3 | 7 - 8 | 2380 | 87 | |
XM_005274483.3 | XP_005274540.2 | 7 - 8 | 2377 | 87 | ||
XM_005274484.2 | XP_005274541.2 | 7 - 8 | 2377 | 87 | ||
XM_011545488.2 | XP_011543790.1 | 7 - 8 | 2377 | 87 | ||
XM_017029399.1 | XP_016884888.1 | 7 - 8 | 2377 | 87 | ||
XM_017029400.1 | XP_016884889.1 | 7 - 8 | 2232 | 87 | ||
XM_017029401.1 | XP_016884890.1 | 7 - 8 | 2377 | 87 | ||
XM_017029402.1 | XP_016884891.1 | 7 - 8 | 2377 | 87 | ||
GenBank mRNA | AL832071.1 | - | 7 - 8 | 2116 | 87 | |
AY331591.1 | - | 8 - 9 | 2264 | 87 | ||
AY331592.1 | - | 8 - 9 | 2264 | 87 | ||
AY371491.1 | - | 3 - 4 | 1717 | 87 | ||
AY437560.1 | - | 3 - 4 | 1741 | 87 | ||
BC032526.1 | - | 4 - 5 | 1918 | 5502039 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000941.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
stimulatory C-type lectin receptor signaling pathway
MyD88-dependent toll-like receptor signaling pathway
I-kappaB kinase/NF-kappaB signaling
JNK cascade
negative regulation of autophagy
Fc-epsilon receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
nucleotide-binding oligomerization domain containing signaling pathway
protein binding
zinc ion binding
stimulatory C-type lectin receptor signaling pathway
MyD88-dependent toll-like receptor signaling pathway
I-kappaB kinase/NF-kappaB signaling
JNK cascade
negative regulation of autophagy
Fc-epsilon receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
nucleotide-binding oligomerization domain containing signaling pathway
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X012538.hg.1 | ENST00000288422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X012538.hg.1 | ENST00000378928.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X012538.hg.1 | ENST00000378930.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X012538.hg.1 | ENST00000378932.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X012538.hg.1 | ENST00000378933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X012538.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X012538.hg.1 | TC0X000941.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000288422.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009339.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | ENST00000378928.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | NM_152787.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | TC0X00009339.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | ENST00000378928.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | ENST00000378930.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | ENST00000378932.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | ENST00000378933.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | ENST00000467136.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00163919.hg.1 | NM_152787.4 | Refseq |
