Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004369.hg.1 |
ENST00000284224.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004369.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004369.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004369.hg.1 |
TC18000430.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000284224.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000284224.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000284224.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000284224.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000284224.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000284224.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000284224.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000284224.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008349.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
ENST00000284224.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
NM_031422.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
TC1800008349.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
ENST00000581714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
ENST00000284224.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
ENST00000580774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
ENST00000618847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
NM_001256316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156944.hg.1 |
NM_031422.5 |
Refseq |