Assay Details
Gene Symbol: DBH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000787.3 | NP_000778.3 | 5 - 6 | 1032 | 110 | |
GenBank mRNA | BC017174.2 | - | 5 - 6 | 1018 | 3163986 | 110 |
CD014125.1 | - | 5 - 6 | 917 | 110 | ||
X13255.1 | - | 5 - 6 | 1013 | 110 | ||
X13256.1 | - | 5 - 6 | 1013 | 110 | ||
Y00096.1 | - | 5 - 6 | 1020 | 110 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000778.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
blood vessel remodeling
response to amphetamine
leukocyte mediated immunity
chemical synaptic transmission
memory
locomotory behavior
visual learning
regulation of cell proliferation
homoiothermy
dopamine catabolic process
norepinephrine biosynthetic process
catecholamine biosynthetic process
glucose homeostasis
fear response
maternal behavior
positive regulation of vasoconstriction
behavioral response to ethanol
response to pain
leukocyte migration
oxidation-reduction process
regulation of extrinsic apoptotic signaling pathway
catalytic activity
dopamine beta-monooxygenase activity
copper ion binding
L-ascorbic acid binding
cytokine production
blood vessel remodeling
response to amphetamine
leukocyte mediated immunity
chemical synaptic transmission
memory
locomotory behavior
visual learning
regulation of cell proliferation
homoiothermy
dopamine catabolic process
norepinephrine biosynthetic process
catecholamine biosynthetic process
glucose homeostasis
fear response
maternal behavior
positive regulation of vasoconstriction
behavioral response to ethanol
response to pain
leukocyte migration
oxidation-reduction process
regulation of extrinsic apoptotic signaling pathway
catalytic activity
dopamine beta-monooxygenase activity
copper ion binding
L-ascorbic acid binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09010097.hg.1 | TC09000778.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09010097.hg.1 | ENST00000371880.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09010097.hg.1 | ENST00000393056.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09010097.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09010089.hg.1 | TC09000778.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09010089.hg.1 | ENST00000371880.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09010089.hg.1 | ENST00000393056.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09010089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09010090.hg.1 | TC09000778.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09010090.hg.1 | ENST00000393056.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09010090.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09010096.hg.1 | TC09000778.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09010096.hg.1 | ENST00000371880.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09010096.hg.1 | ENST00000393056.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09010096.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09010093.hg.1 | TC09000778.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09010093.hg.1 | ENST00000371880.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09010093.hg.1 | ENST00000393056.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09010093.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09010094.hg.1 | TC09000778.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09010094.hg.1 | ENST00000371880.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09010094.hg.1 | ENST00000393056.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09010094.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161941.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161941.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161941.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161941.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161941.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161941.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161941.hg.1 | TC0900009089.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900161944.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161944.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161944.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161944.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161944.hg.1 | TC0900009089.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900161943.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161943.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161943.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161943.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161943.hg.1 | TC0900009089.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900161937.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161937.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161937.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161937.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161937.hg.1 | TC0900009089.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009089.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161936.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161936.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161936.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161936.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161936.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161936.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161936.hg.1 | TC0900009089.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900161940.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161940.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161940.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161940.hg.1 | ENST00000263611.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161940.hg.1 | ENST00000393056.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900161940.hg.1 | NM_000787.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900161940.hg.1 | TC0900009089.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 123258 | - | - |
