Assay Details
Gene Symbol: TRAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190945.1 | NP_001177874.1 | 4 - 5 | 652 | 86 | |
NM_005658.4 | NP_005649.1 | 3 - 4 | 799 | 86 | ||
GenBank mRNA | AK090468.1 | - | 3 - 4 | 799 | 86 | |
AK315476.1 | - | 4 - 5 | 680 | 86 | ||
AL832989.1 | - | 3 - 4 | 2672 | 86 | ||
BC024145.2 | - | 2 - 3 | 305 | 3832475 | 86 | |
BT019408.1 | - | 2 - 3 | 227 | 86 | ||
U19261.1 | - | 2 - 3 | 302 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
